Assay Details
Gene Symbol: Gcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027375.2 | NP_081651.2 | 14 - 15 | 3726 | 87 | |
GenBank mRNA | AB093232.1 | - | 14 - 15 | 3729 | 87 | |
BC023811.1 | - | 9 - 10 | 1113 | 87 | ||
BC034077.1 | - | 3 - 4 | 364 | 87 | ||
BC139007.1 | - | 14 - 15 | 3722 | 8862029 | 87 | |
BC145495.1 | - | 14 - 15 | 3795 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
