Assay Details
Gene Symbol: Gcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027375.2 | NP_081651.2 | 3 - 4 | 182 | 86 | |
GenBank mRNA | AB093232.1 | - | 3 - 4 | 184 | 86 | |
AK011206.1 | - | 3 - 4 | 200 | 86 | ||
AK045701.1 | - | 3 - 4 | 189 | 86 | ||
BC027339.1 | - | 3 - 4 | 164 | 86 | ||
BC139007.1 | - | 3 - 4 | 178 | 8862029 | 86 | |
BC145495.1 | - | 3 - 4 | 251 | 86 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC027339.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
