Assay Details
Gene Symbol: Gcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027375.2 | NP_081651.2 | 8 - 9 | 2978 | 72 | |
GenBank mRNA | AB093232.1 | - | 8 - 9 | 2981 | 72 | |
AK033267.1 | - | 1 - 2 | 1392 | 72 | ||
AK148283.1 | - | 3 - 4 | 193 | 72 | ||
BC023811.1 | - | 3 - 4 | 365 | 72 | ||
BC139007.1 | - | 8 - 9 | 2974 | 8862029 | 72 | |
BC145495.1 | - | 8 - 9 | 3047 | 72 | ||
BY741402.1 | - | 3 - 4 | 193 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
