Assay Details
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175258.4 | NP_780467.2 | 17 - 18 | 2442 | 136 | |
GenBank mRNA | AK143331.1 | - | 5 - 6 | 829 | 136 | |
AK220459.1 | - | 6 - 7 | 845 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
