Assay Details
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033597.1 | NP_001028769.1 | 9 - 10 | 998 | 67 | |
NM_001033598.1 | NP_001028770.1 | 9 - 10 | 967 | 67 | ||
NM_001033599.1 | NP_001028771.1 | 9 - 10 | 1173 | 67 | ||
NM_144823.4 | NP_659072.3 | 9 - 10 | 998 | 67 | ||
GenBank mRNA | AK032479.1 | - | 10 - 11 | 1257 | 67 | |
AK039380.1 | - | 9 - 10 | 1084 | 67 | ||
AK147283.1 | - | 9 - 10 | 968 | 67 | ||
AK147421.1 | - | 9 - 10 | 999 | 67 | ||
AY786360.1 | - | 11 - 12 | 1516 | 67 | ||
AY786361.1 | - | 9 - 10 | 948 | 67 | ||
AY786362.1 | - | 9 - 10 | 1140 | 67 | ||
AY786363.1 | - | 9 - 10 | 993 | 67 | ||
BC016114.1 | - | 9 - 10 | 924 | 4481949 | 67 | |
BC022959.1 | - | 9 - 10 | 998 | 5362660 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
