Assay Details
Gene Symbol: C1qtnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026979.5 | NP_081255.1 | 2 - 3 | 159 | 84 | |
GenBank mRNA | AK007683.1 | - | 2 - 3 | 160 | 84 | |
BC030324.1 | - | 2 - 3 | 160 | 5375090 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007683.1, BC030324.1, NM_026979.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
positive regulation of glycogen biosynthetic process
positive regulation of fatty acid oxidation
positive regulation of glucose import
protein homooligomerization
protein trimerization
protein heterotrimerization
receptor binding
identical protein binding
activation of MAPK activity
positive regulation of glycogen biosynthetic process
positive regulation of fatty acid oxidation
positive regulation of glucose import
protein homooligomerization
protein trimerization
protein heterotrimerization
receptor binding
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27750786 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27750785 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27750784 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P601021 | AK007683.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P601021 | DQ002395.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P601021 | AK007683.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P601021 | DQ002395.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398866 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456625 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762506 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30220 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871005 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34672 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288111 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798080 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23821 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155898 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150943 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179159 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P601021 | AK007683.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P601021 | DQ002395.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P601021 | AK007683.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P601021 | DQ002395.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750703 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750685 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750686 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750680 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750676 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750658 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750697 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750679 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750700 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750692 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750660 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750688 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750668 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27750699 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02310729 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150943 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179159 | - | - |
Whole Mouse Genome G4122A | A_52_P601021 | AK007683.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P601021 | DQ002395.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P601021 | AK007683.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P601021 | DQ002395.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179159 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150943 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09743743 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09743745 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09000241 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09743742 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09338001 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27750786 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27750785 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27750784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27750766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09204295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09416479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09204280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09604974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09571363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09276818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09197275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750663 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09571365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09000242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09204298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09735277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08989563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09223401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09735276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09338001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750668 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09204300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27750701 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10375312 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10375308 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10375310 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10375311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10375309 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431079_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10375307 | - | - |
Mouse Genome 430 2.0 Array | 1431079_at | - | - |
Mouse Genome 430 2.0 Array | 1431079_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69183_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69183_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100003344.mm.1 | NM_026979.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003344.mm.1 | NM_026979.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003344.mm.1 | TC1100000427.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100003345.mm.1 | NM_026979.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003345.mm.1 | NM_026979.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003345.mm.1 | TC1100000427.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100003346.mm.1 | NM_026979.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003346.mm.1 | NM_026979.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003346.mm.1 | TC1100000427.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100003341.mm.1 | NM_026979.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003341.mm.1 | NM_026979.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003341.mm.1 | TC1100000427.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100003347.mm.1 | NM_026979.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003347.mm.1 | NM_026979.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003347.mm.1 | TC1100000427.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100003342.mm.1 | NM_026979.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003342.mm.1 | NM_026979.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100003342.mm.1 | TC1100000427.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431079_PM_at | - | - |
Mouse Expression Set 430 | 1431079_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431079_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431079_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 635291 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232458 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1232458 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1232458 | - | - |
