Assay Details
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019777.3 | NP_062751.2 | 10 - 11 | 1536 | 70 | |
GenBank mRNA | AB016589.1 | - | 8 - 9 | 1224 | 70 | |
AK088580.1 | - | 10 - 11 | 1538 | 70 | ||
AK149911.1 | - | 10 - 11 | 1525 | 70 | ||
AK153046.1 | - | 10 - 11 | 1537 | 70 | ||
AK193494.1 | - | 1 - 2 | 158 | 70 | ||
BC037446.1 | - | 9 - 10 | 1208 | 3980145 | 70 | |
CT010206.1 | - | 8 - 9 | 1190 | 70 | ||
CT010313.1 | - | 8 - 9 | 1118 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
