Assay Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007963.2 | NP_031989.2 | 3 - 4 | 901 | 76 | |
GenBank mRNA | AJ001482.1 | - | 1 - 2 | 57 | 76 | |
AK033517.1 | - | 3 - 4 | 335 | 76 | ||
AK051098.1 | - | 3 - 4 | 758 | 76 | ||
AK085548.1 | - | 3 - 4 | 902 | 76 | ||
BC076620.1 | - | 3 - 4 | 853 | 76 | ||
JQ665270.1 | - | 1 - 2 | 49 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
