Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 21 - 22 | 2469 | 66 | |
NM_001177626.1 | NP_001171097.1 | 21 - 22 | 2507 | 66 | ||
NM_007900.3 | NP_031926.2 | 22 - 23 | 2562 | 66 | ||
GenBank mRNA | AK157718.1 | - | 21 - 22 | 2461 | 66 | |
AK220452.1 | - | 21 - 22 | 2468 | 66 | ||
BC023881.1 | - | 21 - 22 | 2467 | 5354733 | 66 | |
BC025565.1 | - | 22 - 23 | 2536 | 5322744 | 66 | |
BC032155.1 | - | 21 - 22 | 2464 | 5067358 | 66 | |
BC045614.1 | - | 21 - 22 | 2442 | 5059799 | 66 | |
L11316.1 | - | 21 - 22 | 2407 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
