Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 3 - 4 | 368 | 68 | |
NM_001177626.1 | NP_001171097.1 | 3 - 4 | 406 | 68 | ||
GenBank mRNA | AK040174.1 | - | 3 - 4 | 341 | 68 | |
AK157718.1 | - | 3 - 4 | 360 | 68 | ||
AK220452.1 | - | 3 - 4 | 367 | 68 | ||
BC023881.1 | - | 3 - 4 | 366 | 5354733 | 68 | |
BC032155.1 | - | 3 - 4 | 363 | 5067358 | 68 | |
BC045614.1 | - | 3 - 4 | 341 | 5059799 | 68 | |
CN702302.1 | - | 3 - 4 | 368 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
