Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 5 - 6 | 647 | 71 | |
NM_001177626.1 | NP_001171097.1 | 5 - 6 | 685 | 71 | ||
NM_007900.3 | NP_031926.2 | 6 - 7 | 740 | 71 | ||
GenBank mRNA | AK040174.1 | - | 5 - 6 | 620 | 71 | |
AK157718.1 | - | 5 - 6 | 639 | 71 | ||
AK220452.1 | - | 5 - 6 | 646 | 71 | ||
BC023881.1 | - | 5 - 6 | 645 | 5354733 | 71 | |
BC025565.1 | - | 6 - 7 | 714 | 5322744 | 71 | |
BC045614.1 | - | 5 - 6 | 620 | 5059799 | 71 | |
L11316.1 | - | 5 - 6 | 585 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
