Assay Details
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033565.2 | NP_291043.1 | 15 - 16 | 3152 | 133 | |
GenBank mRNA | AF190449.1 | - | 15 - 16 | 3094 | 133 | |
AK053034.1 | - | 3 - 4 | 182 | 133 | ||
BC138999.1 | - | 15 - 16 | 2825 | 8862021 | 133 | |
BC145490.1 | - | 14 - 15 | 2738 | 9054053 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
