Assay Details
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033565.2 | NP_291043.1 | 16 - 17 | 3284 | 86 | |
GenBank mRNA | AF190449.1 | - | 16 - 17 | 3226 | 86 | |
AK033163.1 | - | 6 - 7 | 1104 | 86 | ||
AK053034.1 | - | 4 - 5 | 314 | 86 | ||
BC138999.1 | - | 16 - 17 | 2957 | 8862021 | 86 | |
BC145490.1 | - | 15 - 16 | 2870 | 9054053 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
