Assay Details
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016846.3 | NP_058542.2 | 16 - 17 | 2111 | 80 | |
GenBank mRNA | AK144218.1 | - | 16 - 17 | 2186 | 80 | |
AK169109.1 | - | 16 - 17 | 2163 | 80 | ||
AK169180.1 | - | 16 - 17 | 2164 | 80 | ||
AK173071.1 | - | 6 - 7 | 706 | 80 | ||
BC017678.1 | - | 16 - 17 | 2111 | 4241244 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
