Assay Details
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016846.3 | NP_058542.2 | 1 - 2 | 129 | 88 | |
GenBank mRNA | AK144218.1 | - | 1 - 2 | 204 | 88 | |
AK169109.1 | - | 1 - 2 | 181 | 88 | ||
AK169180.1 | - | 1 - 2 | 182 | 88 | ||
BC017678.1 | - | 1 - 2 | 129 | 4241244 | 88 | |
U14103.1 | - | 1 - 2 | 187 | 88 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK144218.1, AK169109.1, AK169180.1, BC017678.1, NM_016846.3, U14103.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
