Assay Details
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016846.3 | NP_058542.2 | 3 - 4 | 444 | 69 | |
GenBank mRNA | AK144218.1 | - | 3 - 4 | 519 | 69 | |
AK169109.1 | - | 3 - 4 | 496 | 69 | ||
AK169180.1 | - | 3 - 4 | 497 | 69 | ||
BC017678.1 | - | 3 - 4 | 444 | 4241244 | 69 | |
U14103.1 | - | 3 - 4 | 502 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
