Assay Details
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016846.3 | NP_058542.2 | 5 - 6 | 715 | 60 | |
GenBank mRNA | AK144218.1 | - | 5 - 6 | 790 | 60 | |
AK169109.1 | - | 5 - 6 | 767 | 60 | ||
AK169180.1 | - | 5 - 6 | 768 | 60 | ||
BC017678.1 | - | 5 - 6 | 715 | 4241244 | 60 | |
U14103.1 | - | 5 - 6 | 773 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
