Assay Details
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178405.3 | NP_848492.1 | 18 - 19 | 2736 | 85 | |
GenBank mRNA | AK129212.1 | - | 18 - 19 | 2649 | 85 | |
AK137602.1 | - | 18 - 19 | 2672 | 85 | ||
AK147331.1 | - | 18 - 19 | 2668 | 85 | ||
AK161840.1 | - | 18 - 19 | 2668 | 85 | ||
AK164509.1 | - | 18 - 19 | 2672 | 85 | ||
AK168703.1 | - | 18 - 19 | 2668 | 85 | ||
BC025807.1 | - | 7 - 8 | 964 | 85 | ||
BC036127.1 | - | 18 - 19 | 2660 | 4955003 | 85 | |
BC041774.1 | - | 15 - 16 | 2386 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
