Assay Details
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143679.1 | - | 1 - 2 | 534 | 64 | |
AK143680.1 | - | 229 | 64 | |||
AK181422.1 | - | 1 - 2 | 63 | 64 | ||
CK623806.1 | - | 1 - 2 | 261 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK143679.1, AK143680.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
