Assay Details
Gene Symbol: Gm20743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040303.1 | 3 - 4 | 576 | 180 | ||
GenBank mRNA | AK042927.1 | - | 3 - 4 | 565 | 180 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dermatopontin | Dpt |
1810032B19Rik, 5033416F05Rik, EQ-1, Eq1 |
-42.0kb | Sense | Chr. 1 - 164796732 - 164824266 on NCBI Build GRCm38 |
Downstream | chemokine (C motif) ligand 1 | Xcl1 |
AI661682, ATAC, LTN, Lptn, SCM-1, SCM-1a, Scyc1 |
61.4kb | Anti-sense | Chr. 1 - 164931648 - 164935510 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
