Assay Details
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146033.3 | NP_666145.3 | 8 - 9 | 1238 | 87 | |
GenBank mRNA | AK037644.1 | - | 6 - 7 | 3997 | 87 | |
AK077556.1 | - | 8 - 9 | 1209 | 87 | ||
AK164434.1 | - | 8 - 9 | 1140 | 87 | ||
BC024959.1 | - | 8 - 9 | 1195 | 4946879 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | AK164434.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | BF453918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | NM_146033.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | AK077556.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | AK164434.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | BF453918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | NM_146033.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P256957 | AK077556.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | AK164434.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | BC012275.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | BC024959.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | NM_146033.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | AK037644.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | AK164434.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | BC012275.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | BC024959.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | NM_146033.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684642 | AK037644.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694845 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366216 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412198 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92408 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77665 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177208 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112078 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | AK164434.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | BC012275.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | BC024959.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | NM_146033.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | AK037644.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | AK164434.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | BC012275.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | BC024959.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | NM_146033.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684642 | AK037644.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | AK164434.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | BF453918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | NM_146033.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | AK077556.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | AK164434.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | BF453918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | NM_146033.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P256957 | AK077556.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177208 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07588442 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07588400 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07588426 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112078 | - | - |
Whole Mouse Genome G4122A | A_52_P684642 | AK164434.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | BC012275.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | BC024959.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | NM_146033.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | AK037644.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | AK164434.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | BC012275.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | BC024959.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | NM_146033.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684642 | AK037644.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | AK164434.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | BF453918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | NM_146033.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | AK077556.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | AK164434.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | BF453918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | NM_146033.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P256957 | AK077556.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127247 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112078 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28356973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28357015 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10395383 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395381 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395382 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395380 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395385 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395388 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395386 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395387 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395378 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395379 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395377 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425936_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451724_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425482_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10395376 | - | - |
Mouse Genome 430 2.0 Array | 1451724_at | - | - |
Mouse Genome 430 2.0 Array | 1425936_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1425482_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1451724_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1425936_a_at | - | - |
Mouse Genome 430 2.0 Array | 1425482_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217473_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217473_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200002399.mm.1 | NM_146033.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002399.mm.1 | NM_146033.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002399.mm.1 | TC1200000319.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200002400.mm.1 | NM_146033.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002400.mm.1 | NM_146033.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002400.mm.1 | TC1200000319.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200002397.mm.1 | NM_146033.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002397.mm.1 | NM_146033.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002397.mm.1 | TC1200000319.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200002398.mm.1 | NM_146033.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002398.mm.1 | NM_146033.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200002398.mm.1 | TC1200000319.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451724_at | - | - |
Mouse Expression Set 430 | 1451724_PM_at | - | - |
Mouse Expression Set 430 | 1425936_PM_a_at | - | - |
Mouse Expression Set 430 | 1425482_PM_s_at | - | - |
Mouse Expression Set 430 | 1425936_a_at | - | - |
Mouse Expression Set 430 | 1425482_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1425936_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1425482_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1425936_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451724_at | - | - |
Mouse Genome 430A 2.0 Array | 1451724_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425482_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425936_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425482_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451724_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425482_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425936_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451724_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 874726 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2230440 | - | - |
MouseWG-6 v2 Expression BeadChip | 780600 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2230440 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101500731 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002230440 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663741 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255153 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2663741 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255153 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1228261 | - | - |
mouse-6 v1.1 (Illumina) | scl43280.11_443-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101500731 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2230440 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl43280.11_443-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230440 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101500731 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2599514 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2663741 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2663741 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228261 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl43280.11_443-S | - | - |
