Assay Details
Gene Symbol: Capn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011796.2 | NP_035926.2 | 8 - 9 | 1931 | 66 | |
GenBank mRNA | AF126867.1 | - | 8 - 9 | 1489 | 66 | |
AK088207.1 | - | 8 - 9 | 1565 | 66 | ||
BC005681.1 | - | 8 - 9 | 1575 | 3154780 | 66 | |
BC010969.1 | - | 8 - 9 | 1575 | 4159612 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
