Assay Details
Gene Symbol: Naa15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053089.3 | NP_444319.3 | 5 - 6 | 847 | 165 | |
GenBank mRNA | AB221683.1 | - | 5 - 6 | 540 | 165 | |
AF510858.1 | - | 5 - 6 | 737 | 165 | ||
BC050017.1 | - | 5 - 6 | 738 | 30020858 | 165 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
