Assay Details
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198111.2 | NP_932779.2 | 8 - 9 | 2967 | 95 | |
GenBank mRNA | AK147248.1 | - | 8 - 9 | 2968 | 95 | |
AK147371.1 | - | 8 - 9 | 2968 | 95 | ||
BC057114.1 | - | 7 - 8 | 2917 | 6405025 | 95 | |
DQ233645.1 | - | 8 - 9 | 3023 | 95 | ||
DQ233646.1 | - | 7 - 8 | 2817 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
