Assay Details
Gene Symbol: Sec23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009147.2 | NP_033173.2 | 15 - 16 | 1937 | 135 | |
GenBank mRNA | AK028152.1 | - | 15 - 16 | 1825 | 135 | |
AY082671.1 | - | 15 - 16 | 1811 | 135 | ||
BC034610.1 | - | 15 - 16 | 1937 | 5364796 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
