Assay Details
Gene Symbol: Sec23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009147.2 | NP_033173.2 | 5 - 6 | 805 | 74 | |
GenBank mRNA | AK028152.1 | - | 5 - 6 | 693 | 74 | |
AY082671.1 | - | 5 - 6 | 679 | 74 | ||
BC034610.1 | - | 5 - 6 | 805 | 5364796 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
