Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 13 - 14 | 2265 | 153 | ||
GenBank mRNA | AK006249.1 | - | 11 - 12 | 1531 | 153 | |
AY461596.1 | - | 13 - 14 | 2283 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
