Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 4 - 5 | 984 | 82 | ||
GenBank mRNA | AK006249.1 | - | 2 - 3 | 250 | 82 | |
AK016463.1 | - | 4 - 5 | 996 | 82 | ||
AY461596.1 | - | 4 - 5 | 1002 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
