Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 6 - 7 | 1216 | 129 | ||
GenBank mRNA | AK006249.1 | - | 4 - 5 | 482 | 129 | |
AK016463.1 | - | 6 - 7 | 1228 | 129 | ||
AY461596.1 | - | 6 - 7 | 1234 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
