Assay Details
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023644.4 | NP_076133.3 | 10 - 11 | 1197 | 72 | |
GenBank mRNA | AF310338.1 | - | 10 - 11 | 1118 | 72 | |
AK007782.1 | - | 10 - 11 | 1150 | 72 | ||
AK031072.1 | - | 10 - 11 | 1158 | 72 | ||
AK032203.1 | - | 10 - 11 | 1140 | 72 | ||
AK160184.1 | - | 8 - 9 | 914 | 72 | ||
AK168589.1 | - | 10 - 11 | 1086 | 72 | ||
AK199182.1 | - | 1 - 2 | 144 | 72 | ||
BC021382.1 | - | 10 - 11 | 5042993 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
