Assay Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164233.1 | NP_001157705.1 | 11 - 12 | 1892 | 131 | |
NM_027584.1 | NP_081860.1 | 11 - 12 | 1892 | 131 | ||
GenBank mRNA | AK014872.1 | - | 11 - 12 | 1893 | 131 | |
AK133206.1 | - | 12 - 13 | 1981 | 131 | ||
BC112423.1 | - | 11 - 12 | 1862 | 40062055 | 131 | |
BC113131.1 | - | 11 - 12 | 1862 | 40062053 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | AK133206.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | BC112423.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | BC113131.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | NM_027584.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | AK014872.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | AK133206.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | BC112423.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | BC113131.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | NM_027584.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430718 | AK014872.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158826 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76032 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777726 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508553 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75746 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147848 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143532 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | AK133206.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | BC112423.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | BC113131.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | NM_027584.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | AK014872.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | AK133206.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | BC112423.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | BC113131.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | NM_027584.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430718 | AK014872.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143532 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04201927 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00142424 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04201888 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04201993 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04201906 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147848 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04201879 | - | - |
Whole Mouse Genome G4122A | A_51_P430718 | AK133206.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | BC112423.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | BC113131.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | NM_027584.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | AK014872.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | AK133206.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | BC112423.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | BC113131.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | NM_027584.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P430718 | AK014872.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04201906 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143532 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00142424 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147848 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476097 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00701871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00369548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00340472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475993 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476002 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20476018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20475973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01130655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00545024 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10348954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348953 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348957 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348952 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348959 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348949 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348950 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348960 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348951 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431449_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10348947 | - | - |
Mouse Genome 430 2.0 Array | 1431449_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431449_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70866_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70866_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100008238.mm.1 | TC0100000977.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008238.mm.1 | NM_001164233.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008238.mm.1 | NM_001164233.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008238.mm.1 | NM_027584.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008238.mm.1 | NM_027584.1 | Refseq |
Mouse Expression Set 430 | 1431449_at | - | - |
Mouse Expression Set 430 | 1431449_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431449_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431449_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6290180 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693887 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2693887 | - | - |
