Assay Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027584.1 | NP_081860.1 | 12 - 13 | 2022 | 95 | |
GenBank mRNA | AK014872.1 | - | 12 - 13 | 2023 | 95 | |
AK133206.1 | - | 13 - 14 | 2111 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
