Assay Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164233.1 | NP_001157705.1 | 5 - 6 | 1043 | 143 | |
NM_027584.1 | NP_081860.1 | 5 - 6 | 1043 | 143 | ||
GenBank mRNA | AK014872.1 | - | 5 - 6 | 1044 | 143 | |
BC112423.1 | - | 5 - 6 | 1013 | 40062055 | 143 | |
BC113131.1 | - | 5 - 6 | 1013 | 40062053 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
