Assay Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164233.1 | NP_001157705.1 | 7 - 8 | 1293 | 157 | |
NM_027584.1 | NP_081860.1 | 7 - 8 | 1293 | 157 | ||
GenBank mRNA | AK014872.1 | - | 7 - 8 | 1294 | 157 | |
AK133206.1 | - | 8 - 9 | 1382 | 157 | ||
BC112423.1 | - | 7 - 8 | 1263 | 40062055 | 157 | |
BC113131.1 | - | 7 - 8 | 1263 | 40062053 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
