Assay Details
Gene Symbol: Pcdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098171.1 | NP_001091641.1 | 4 - 5 | 3959 | 62 | |
GenBank mRNA | AF334801.1 | - | 4 - 5 | 3195 | 62 | |
AK018296.1 | - | 1 - 2 | 163 | 62 | ||
AK034005.1 | - | 4 - 5 | 834 | 62 | ||
AK036195.1 | - | 4 - 5 | 2302 | 62 | ||
AK039226.1 | - | 4 - 5 | 1305 | 62 | ||
AK122503.1 | - | 4 - 5 | 3928 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
