Assay Details
Gene Symbol: Dcaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028759.1 | NP_083035.1 | 7 - 8 | 1140 | 84 | |
GenBank mRNA | AK004618.1 | - | 7 - 8 | 1003 | 84 | |
BC058980.1 | - | 5 - 6 | 703 | 84 | ||
BC084732.1 | - | 5 - 6 | 703 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
