Assay Details
Gene Symbol: Fat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029988.2 | NP_001025159.1 | 9 - 10 | 8865 | 87 | |
GenBank mRNA | AK173031.1 | - | 1 - 2 | 763 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
