Assay Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172644.3 | NP_766232.1 | 1 - 2 | 647 | 76 | |
GenBank mRNA | AK030952.1 | - | 1 - 2 | 636 | 76 | |
AK037337.1 | - | 1 - 2 | 666 | 76 | ||
AK048454.1 | - | 1 - 2 | 624 | 76 | ||
AK138809.1 | - | 1 - 2 | 648 | 76 | ||
AK154133.1 | - | 1 - 2 | 676 | 76 | ||
AK154348.1 | - | 1 - 2 | 676 | 76 | ||
AK180299.1 | - | 1 - 2 | 65 | 76 | ||
BC057943.1 | - | 1 - 2 | 671 | 76 | ||
BC076606.1 | - | 1 - 2 | 639 | 30625515 | 76 | |
BC089191.1 | - | 1 - 2 | 621 | 30473852 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
