Assay Details
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144942.4 | NP_659191.1 | 6 - 7 | 732 | 58 | |
GenBank mRNA | AK005015.1 | - | 6 - 7 | 733 | 58 | |
AY033912.1 | - | 6 - 7 | 649 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
