Assay Details
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080925.1 | NP_001074394.1 | 11 - 12 | 2024 | 122 | |
GenBank mRNA | BC080279.1 | - | 10 - 11 | 1092 | 122 | |
BC130266.1 | - | 12 - 13 | 1300 | 40143028 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
