Assay Details
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080925.1 | NP_001074394.1 | 13 - 14 | 2180 | 75 | |
GenBank mRNA | AK045359.1 | - | 1 - 2 | 101 | 75 | |
BC080279.1 | - | 12 - 13 | 1248 | 75 | ||
BC130266.1 | - | 14 - 15 | 1456 | 40143028 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
