Assay Details
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080925.1 | NP_001074394.1 | 1 - 2 | 884 | 116 | |
GenBank mRNA | BC130266.1 | - | 2 - 3 | 160 | 40143028 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC130266.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
