Assay Details
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080925.1 | NP_001074394.1 | 2 - 3 | 965 | 72 | |
GenBank mRNA | BC130266.1 | - | 3 - 4 | 241 | 40143028 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC130266.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
