Assay Details
Gene Symbol: Myo1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177390.3 | NP_796364.2 | 5 - 6 | 851 | 77 | |
GenBank mRNA | AK037051.1 | - | 5 - 6 | 830 | 78 | |
AK049374.1 | - | 5 - 6 | 852 | 77 | ||
BC150765.1 | - | 5 - 6 | 717 | 9087676 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
