Assay Details
Gene Symbol: Fmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011711.1 | NP_035841.1 | 18 - 19 | 2401 | 91 | |
GenBank mRNA | AK129473.1 | - | 18 - 19 | 2320 | 91 | |
AK143308.1 | - | 18 - 19 | 2301 | 91 | ||
AK155331.1 | - | 18 - 19 | 2418 | 91 | ||
AK163361.1 | - | 18 - 19 | 2388 | 91 | ||
AK170744.1 | - | 17 - 18 | 2231 | 91 | ||
BC023797.1 | - | 17 - 18 | 2121 | 91 | ||
BC027845.1 | - | 2 - 3 | 378 | 91 | ||
BC085094.1 | - | 18 - 19 | 2296 | 91 | ||
BC131961.1 | - | 18 - 19 | 2277 | 40130238 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
