Assay Details
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205353.1 | NP_001192282.1 | 8 - 9 | 1009 | 73 | |
NM_172611.4 | NP_766199.1 | 8 - 9 | 911 | 73 | ||
GenBank mRNA | AK036832.1 | - | 8 - 9 | 900 | 73 | |
AK129209.1 | - | 8 - 9 | 813 | 73 | ||
AK142266.1 | - | 8 - 9 | 864 | 73 | ||
BC021523.1 | - | 4 - 5 | 285 | 73 | ||
BC085148.1 | - | 8 - 9 | 967 | 73 | ||
BC139228.1 | - | 8 - 9 | 900 | 8862250 | 73 | |
BC139229.1 | - | 8 - 9 | 900 | 8862251 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
