Assay Details
Gene Symbol: Usp32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029934.1 | NP_001025105.1 | 1 - 2 | 248 | 79 | |
GenBank mRNA | AK148221.1 | - | 1 - 2 | 249 | 79 | |
AK170234.1 | - | 1 - 2 | 247 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
