Assay Details
Gene Symbol: C030037D09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038058.1 | 3 - 4 | 644 | 74 | ||
GenBank mRNA | AK021127.1 | - | 3 - 4 | 452 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | musashi RNA-binding protein 2 | Msi2 |
1700105C15Rik, AI451722, AI563628, AW489193, Msi2h |
-9.3kb | Anti-sense | Chr. 11 - 88339382 - 88718267 on NCBI Build GRCm38 |
Downstream | A kinase (PRKA) anchor protein 1 | Akap1 |
AKAP121, AKAP84, Akap, C76494, C81186, DAKAP1, S-AKAP84 |
102.2kb | Anti-sense | Chr. 11 - 88830792 - 88864586 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
