Assay Details
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027427.2 | NP_081703.1 | 13 - 14 | 1175 | 79 | |
GenBank mRNA | AK051568.1 | - | 13 - 14 | 1176 | 79 | |
AK146331.1 | - | 13 - 14 | 1138 | 79 | ||
AK166550.1 | - | 13 - 14 | 1168 | 79 | ||
BC043716.1 | - | 7 - 8 | 503 | 79 | ||
BC137591.1 | - | 13 - 14 | 1149 | 8860610 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
