Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 19 - 20 | 4591 | 139 | |
GenBank mRNA | AY557610.1 | - | 19 - 20 | 4582 | 139 | |
BC046463.1 | - | 15 - 16 | 1888 | 139 | ||
BC141180.1 | - | 18 - 19 | 4087 | 9055792 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
